Learn More
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Melting Point | -90°C |
| Color | Colorless |
| Density | 0.785g/mL at 25°C |
| Quantity | 2.5 L |
| Explosion Limit | 0.02 |
| Flash Point | 12°C (53.6°F) |
| Ignition Point | 750°F |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | n20/D 1.377(lit.) |
| Show More |
Safety and Handling
| Hazard Statement | P210-P305 + P351 + P338-P370 + P378-P403 + P235 |
| Precautionary Statement | H225-H319-H336 |
| DOTInformation | Class 3 - PG 2 - Isopropanol |
| EINECSNumber | 200-661-7 |
| RTECSNumber | NT8050000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.