Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, ACS Grade, Fisher Chemical™

Catalog No. A416SK4 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Aluminum Can
Glass Bottle
HDPE Bottle
Poly Bottle
Safe-Cote™
Steel Drum
Steel Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A416SK-4 4 L Safe-Cote™
A416-1 1 L HDPE Bottle
A416-4 4 L Glass Bottle
A416P-4 4 L Poly Bottle
A416-20 20 L Steel Pail
A416-200 200 L Steel Drum
A416-500 500 mL HDPE Bottle
A416S-4 4 L Aluminum Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. A416SK-4

In addition to meeting or exceeding the latest ACS specifications, are analyzed for more than 16 metals; Actual lot of analysis on label

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH 7
Melting Point -89.5°C
Color Colorless
Density 0.785g/cm³
Quantity 4 L
Flash Point 12°C (53.6°F)
Identification Pass Test
Packaging Safe-Cote™
Physical Form Liquid
Refractive Index 1.3740 to 1.3760 (at 25°C)
Residue after Evaporation 0.001% max.
Specific Gravity 0.785
Assay Percent Range ≥99.5 %
Vapor Pressure 43mmHg at 20°C
Linear Formula (CH3)2CHOH
Viscosity 2.27 mPa/s at 20°C
UN Number 1219
Water 0.2% max.
Boiling Point 81°C
Formula Weight 60.1
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material 2-Propanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.