Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol (Laboratory), Fisher Chemical™

Catalog No. A426F1GAL Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
4 L
20 L
200 L
Packaging:
Aluminum Can
Poly Bottle
Steel Drum
Steel Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A426F-1GAL 1 gal. Poly Bottle
A426S-4 4 L Aluminum Can
A426P-4 4 L Poly Bottle
A426S-20 20 L Steel Pail
A426S-200 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A426F-1GAL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specially filtered for use in tissue processors

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH 7
Melting Point -89.5°C
Color Colorless
Density 0.785g/cm³
Quantity 1 gal.
Identification Pass Test
Packaging Poly Bottle
Physical Form Liquid
Vapor Pressure 43mmHg at 20°C
Linear Formula (CH3)2CHOH
Viscosity 2.27 mPa/s at 20°C
Boiling Point 81°C
Formula Weight 60.1
Grade Laboratory
Chemical Name or Material 2-Propanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.