Learn More
Sigma Aldrich Fine Chemicals Biosciences 2 Propanol HPLC Plus for H
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 6504471L/DEL
Encompass
2-Propanol (Isopropanol) is a secondary alcohol. It has been tested as a substitute of the fuel for use in various fuel cells. CuO powder dissolved in 2-propanol has been used for the laser ablation assisted synthesis of Cu colloids. Suspension of 2-propanol with Zn(NO3)2 has been employed for the fabrication of titanium dioxide-coated stainless steel (P25-TiO2/SS) photoanode coated with uniformly thick layer of P25-TiO2. This photoanode was used for the electrochemical photocatalytic (ECPC) degradation process. Preparation of methyl isobutyl ketone (MIBK) using 2-propanol as the precursor has been reported with yield as high as 25%. Solubility of benzoic acid has been investigated in 2-propanol by gravimetric method in the temperature range 277-346K.
Catalog No. 50-290-6766