Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, Laboratory Reagent, ≥99.5%, Solstice

Catalog No. 60044667 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-667 20 L Stainless Steel Can
60-044-666 2.5 L Glass Bottle
60-044-665 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-044-667 Supplier Solstice Supplier No. 10982720L

Laboratory Reagent,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Color Colorless
Density 0.785g/mL at 25°C
Quantity 20 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Packaging Stainless Steel Can
Physical Form Liquid
Refractive Index n20/D 1.377(lit.)
Vapor Pressure 33mmHg (20°C), 44mmHg (25°C)
Linear Formula (CH3)2CHOH
UN Number UN1219
Beilstein 635639
Boiling Point 77°C to 83°C (1013 hPa)
Solubility Information Soluble in water
Formula Weight 60.1g/mol
Percent Purity ≥99.5%
Grade Laboratory Reagent
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
Precautionary Statement H225-H319-H336
DOTInformation Class 3 - PG 2 - Isopropanol
EINECSNumber 200-661-7
RTECSNumber NT8050000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.