Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP, Solstice

Catalog No. 60047432 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-432 2.5 L Poly Bottle
60-047-431 1 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-432 Supplier Solstice Supplier No. 722992.5L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100.0000%
Density 0.784 - 0.786 g/cm3
Quantity 2.5 L
Flash Point 12°C
Packaging Poly Bottle
UN Number UN1219
Beilstein 635639
Boiling Point 77°C to 83°C
Formula Weight 60.11
Percent Purity >99.9%
Grade ACS
Chemical Name or Material 2-Propanol, puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP
Show More Show Less
ShelfLife 1080 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.