Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran inhibitor
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 3486512X100ML
Encompass
Tetrahydrofuran (THf) is widely employed as a solvent. It constitutes the key fragment of various natural products (polyether antibiotics). THf forms a double hydrate with hydrogen sulfide. Crystal structure of this double hydrate has been investigated by three-dimensional single-crystal studies. Butane-1 4-diol is formed as an intermediate during the synthesis of THf. Hot THf is useful for the disSOLUTIONon of polyvinylidene chloride (PVDV).
Catalog No. 50-290-3989
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.