Learn More
Sigma Aldrich Fine Chemicals Biosciences SMCTM Human Total Tau High
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 03018500
Encompass
The human Tau family of microtubule-associated neuronal proteins has been rigorously studied in the context of normal and aberrant neurobiology. While typically involved in the stability of axonal microtubules Tau proteins can also play a role in neurodegenerative diseases such as Alzheimers Disease (AD) and frontotemporal dementia (fTD) if they aberrantly misfold and accumulate in the brain. There is significant interest in studying the level of Tau proteins in both the central nervous system as well as in circulation and the new SMC(TM) Human Total-Tau High Sensitivity Immunoassay Kit enables researchers to take advantage of the Single Molecule Counting ultrasensitive immunoassay technology to perform accurately and sensitive measurements in human cerebrospinal fluid (CSf) plasma and serum samples toward making important discoveries about the role of Tau protein in neurodegenerative disease mechanisms.The SMC(TM) Human Total Tau High Sensitivity Immunoassay Kit contains all reag
Catalog No. 50-290-5723
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.