Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Propene-1-thiol

Catalog No. 06030100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-030-100ML 100 mL
06-030-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-030-100ML Supplier Sigma Aldrich Supplier No. 06030100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Propene-1-thiol

Specifications

CAS 870-23-5
Density 0.898 g/mL (at 25°C (literature))
Boiling Point 67°C to 68°C (lit.)
Molecular Formula C3H6S
Linear Formula CH2=CHCH2SH
Refractive Index n20/D 1.4765 (literature)
MDL Number MFCD00004894
Quantity 100 mL
Synonym Allyl mercaptan
Molecular Weight (g/mol) 74.14
Percent Purity ∼60% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.