Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propionylthiophene 98.0+%, TCI America™
SDP

Catalog No. P1097250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1097250G 250 g
P109725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1097250G Supplier TCI America Supplier No. P1097250G

Chemical Identifiers

CAS 13679-75-9
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00005446
InChI Key MFPZQZZWAMAHOY-UHFFFAOYSA-N
Synonym 1-2-thienyl-1-propanone, 2-propionylthiophene, 1-thiophen-2-yl propan-1-one, 1-2-thienyl propan-1-one, ethyl 2-thienyl ketone, 2-propanoylthiophene, 1-propanone, 1-2-thienyl, unii-ny658tvp5m, ethyl thienyl ketone, ethyl-2-thienyl ketone
PubChem CID 26179
IUPAC Name 1-thiophen-2-ylpropan-1-one
SMILES CCC(=O)C1=CC=CS1

Specifications

Boiling Point 228°C
Quantity 250 g
Formula Weight 140.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propionylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.