Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propylpyridine 98.0+%, TCI America™
SDP

Catalog No. P033625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P033625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P033625ML Supplier TCI America Supplier No. P033625ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 622-39-9
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00006368
InChI Key OIALIKXMLIAOSN-UHFFFAOYSA-N
Synonym 2-n-propylpyridine, conyrine, conyrin, pyridine, 2-propyl, 1-2-pyridyl propane, 2-propyl-pyridine, unii-3n6ax0b7pa, 2-propyl pyridine, pyridine, propyl, pyridine, 1-propyl
PubChem CID 69320
IUPAC Name 2-propylpyridine
SMILES CCCC1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 167°C
Quantity 25 mL
UN Number 1993
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Propylpyridine
EINECSNumber (9)-1272
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.