Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propyltoluene 98.0+%, TCI America™
SDP

Catalog No. P074725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P074725ML 25 mL
P07475ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P074725ML Supplier TCI America Supplier No. P074725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1074-17-5
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00059246
InChI Key YQZBFMJOASEONC-UHFFFAOYSA-N
Synonym 1-Methyl-2-propylbenzene
PubChem CID 14091
IUPAC Name 1-methyl-2-propylbenzene
SMILES CCCC1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 185°C
Quantity 25 mL
UN Number 3295
Formula Weight 134.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propyltoluene
EINECSNumber (3)-0015
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.