Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyridinecarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. P0425100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0425100G 100 g
P042525G 25 g
P0425500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0425100G Supplier TCI America Supplier No. P0425100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1121-60-4
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.112
MDL Number MFCD00006290
InChI Key CSDSSGBPEUDDEE-UHFFFAOYSA-N
Synonym 2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde
PubChem CID 14273
ChEBI CHEBI:73012
IUPAC Name pyridine-2-carbaldehyde
SMILES C1=CC=NC(=C1)C=O

Specifications

Melting Point -21°C
Color Yellow
Boiling Point 181°C
Quantity 100 g
UN Number 2810
Formula Weight 107.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pyridinecarboxaldehyde
EINECSNumber (5)-6697
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.