Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Quinolinecarboxaldehyde 97.0+%, TCI America™
SDP

Catalog No. Q00641G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00641G 1 g
Q00645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00641G Supplier TCI America Supplier No. Q00641G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5470-96-2
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
MDL Number MFCD00075032
InChI Key WPYJKGWLDJECQD-UHFFFAOYSA-N
Synonym 2-quinolinecarboxaldehyde, quinoline-2-carboxaldehyde, 2-quinolinecarbaldehyde, 2-formylquinoline, quinaldaldehyde, quinolinecarboxaldehyde, quinoline aldehyde, quinoline-2-aldehyde, zlchem 599, pubchem2277
PubChem CID 79619
IUPAC Name quinoline-2-carbaldehyde
SMILES O=CC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 70°C
Quantity 1 g
Formula Weight 157.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Quinolinecarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.