Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(tert-Butoxycarbonylamino)isobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. B332325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B332325G 25 g
B33235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B332325G Supplier TCI America Supplier No. B332325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30992-29-1
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00042973
InChI Key MFNXWZGIFWJHMI-UHFFFAOYSA-N
Synonym boc-aib-oh, boc-alpha-methylalanine, 2-boc-amino isobutyric acid, 2-tert-butoxycarbonyl amino-2-methylpropanoic acid, boc-2-aminoisobutyric acid, n-boc-2-aminoisobutyric acid, n-boc-alpha-methylalanine, n-boc-2-amino-2-methylpropanoic acid, 2-tert-butoxy carbonyl amino-2-methylpropanoic acid, 2-tert-butoxycarbonylamino isobutyric acid
PubChem CID 2733845
IUPAC Name 2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(C)(C)C(=O)O

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 25 g
Formula Weight 203.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(tert-Butoxycarbonylamino)isobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.