Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(tert-Butoxycarbonyloxyimino)-2-phenylacetonitrile 98.0+%, TCI America™
SDP

Catalog No. B098825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B098825G 25 g
B09885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B098825G Supplier TCI America Supplier No. B098825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58632-95-4
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.266
MDL Number MFCD00001863
InChI Key QQWYQAQQADNEIC-PTNGSMBKSA-N
Synonym 2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile, 2-boc-oxyimino-2-phenylacetonitrile, boc-on, ccris 2601, tert-butyl alpha-cyanobenzylaminyl carbonate, 2-tert-butoxycarbonyloximino-2-phenylacetonitrile, carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester, n-tert-butoxycarbonyl oxy benzimidoyl cyanide, benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino
PubChem CID 5868400
IUPAC Name tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate
SMILES CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1

Specifications

Melting Point 88°C
Color White
Quantity 25 g
UN Number 3439
Formula Weight 246.27
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(tert-Butoxycarbonyloxyimino)-2-phenylacetonitrile
EINECSNumber (3)-3524
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.