Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. I084225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I084225G 25 g
I08425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I084225G Supplier TCI America Supplier No. I084225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28159-98-0
Molecular Formula C11H19N5S
Molecular Weight (g/mol) 253.368
MDL Number MFCD01863779
InChI Key HDHLIWCXDDZUFH-UHFFFAOYSA-N
Synonym Cybutryne
PubChem CID 91590
ChEBI CHEBI:5962
IUPAC Name 2-N-tert-butyl-4-N-cyclopropyl-6-methylsulfanyl-1,3,5-triazine-2,4-diamine
SMILES CC(C)(C)NC1=NC(=NC(=N1)NC2CC2)SC

Specifications

Melting Point 128°C
Color White
Quantity 25 g
UN Number 3077
Formula Weight 253.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-1,3,5-triazine
EINECSNumber (5)-6110
RTECSNumber XY5850675
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.