Learn More
Sigma Organic Chemistry 2- tert-Butylcarbonylam | 1G | 146140-95-6 | MFCD01114645
Supplier: Sigma Organic Chemistry BML000191G

2- tert-Butylcarbonylam , 1G
About this Item:
CAS #: 146140-95-6
MDL #: MFCD01114645
Molecular Weight: 221.06
UNSPSC Code: 12352200
Chemical Formula: C11H16BNO3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.