Learn More
Sigma Organic Chemistry 2- tert-Butylsulfanyl p 2 | 25mg | MFCD01830291
Supplier: Sigma Organic Chemistry CDS01317125MG

2- tert-Butylsulfanyl p 2, 25mg
About this Item:
MDL #: MFCD01830291
Molecular Weight: 224.32
UNSPSC Code: 12352200
Chemical Formula: C12H16O2S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.