Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Tetrahydro-2H-pyran-2-yloxy)ethanol

Catalog No. 8747725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
87-477-25ML 25 mL
87-477-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 87-477-25ML Supplier Sigma Aldrich Supplier No. 8747725ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(Tetrahydro-2H-pyran-2-yloxy)ethanol

Specifications

CAS 2162-31-4
Density 1.077 g/mL (at 20°C (literature))
Boiling Point 95°C (22 mmHg)
Molecular Formula C7H14O3
Refractive Index n20/D 1.457
Linear Formula C7H14O3
MDL Number MFCD00185588
Quantity 25 mL
Synonym 2-(2-Hydroxyethoxy)tetrahydropyran
Molecular Weight (g/mol) 146.18
Percent Purity ≥98.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.