Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thenoyl Chloride 99.0+%, TCI America™
SDP

Catalog No. T0177100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0177100G 100 g
T017725G 25 g
T0177500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0177100G Supplier TCI America Supplier No. T0177100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5271-67-0
Molecular Formula C5H3ClOS
Molecular Weight (g/mol) 146.59
MDL Number MFCD00005428
InChI Key QIQITDHWZYEEPA-UHFFFAOYSA-N
Synonym 2-thiophenecarbonyl chloride, 2-thenoyl chloride, 2-thienylcarbonyl chloride, 2-thiophenecarbonylchloride, thiophenecarbonyl chloride, 2-thiophene carbonyl chloride, thiophene-2-carboxylic acid chloride, thenoyl chloride, thienoyl chloride, 2-thenoylchloride
PubChem CID 78928
IUPAC Name thiophene-2-carbonyl chloride
SMILES ClC(=O)C1=CC=CS1

Specifications

Boiling Point 190°C
Quantity 100 g
UN Number 3265
Formula Weight 146.59
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Thenoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.