Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Thenoyltrifluoroacetone

Catalog No. T27006100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27006100G 100 g
T2700625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27006100G Supplier Sigma Aldrich Supplier No. T27006100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Thenoyltrifluoroacetone

Specifications

CAS 326-91-0
Melting Point 40°C to 44°C (lit.)
Boiling Point 96°C to 98°C (8 mmHg)
Molecular Formula C8H5F3O2S
Linear Formula C8H5F3O2S
MDL Number MFCD00005445
Quantity 100 g
Synonym 4,4,4-Trifluoro-1-(2-thienyl)-1,3-butanedione; TTA (extractant); TTB; TTFA
Molecular Weight (g/mol) 222.18
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.