Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiopheneacetamide, 98%

Catalog No. AAL1922114
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1922114 25 g
AAL1922106 5 g
AAL1922122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1922114 Supplier Thermo Scientific Chemicals Supplier No. L1922114
Add to Cart

CAS: 4461-29-4 | C6H7NOS | 141.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4461-29-4
Molecular Formula C6H7NOS
Molecular Weight (g/mol) 141.188
MDL Number MFCD00017556
InChI Key UUPZTFTUZUQRQT-UHFFFAOYSA-N
Synonym 2-thiophen-2-yl acetamide, thiophene-2-acetamide, 2-thiopheneacetamide, 2-2-thienyl acetamide, thiophen-2-acetamide, thiophen-2-yl-acetamide, 2-thienylacetamide, thiolacet-amide, 2-thienyl acetamide, acmc-1ajdb
PubChem CID 78208
IUPAC Name 2-thiophen-2-ylacetamide
SMILES C1=CSC(=C1)CC(=O)N

Specifications

Melting Point 145°C to 148°C
Quantity 25 g
Beilstein 114558
Formula Weight 141.19
Percent Purity 98%
Chemical Name or Material 2-Thiopheneacetamide
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
EINECSNumber 224-714-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.