Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiopheneacrylic Acid 98.0+%, TCI America™
SDP

Catalog No. T113825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T113825G 25 g
T11385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T113825G Supplier TCI America Supplier No. T113825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15690-25-2
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.183
MDL Number MFCD00005455
InChI Key KKMZQOIASVGJQE-ONEGZZNKSA-N
Synonym 3-2-thienyl acrylic acid, 3-thiophen-2-yl acrylic acid, 2-thiopheneacrylic acid, 2e-3-thiophen-2-yl prop-2-enoic acid, 2e-3-2-thienyl acrylic acid, 2-thienylacrylic acid, e-3-thiophen-2-yl acrylic acid, 2-thiopheneacrylic acid, e, unii-ino5aty68d, 2-propenoic acid, 3-2-thienyl
PubChem CID 735981
IUPAC Name (E)-3-thiophen-2-ylprop-2-enoic acid
SMILES C1=CSC(=C1)C=CC(=O)O

Specifications

Melting Point 146°C
Quantity 25 g
Formula Weight 154.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiopheneacrylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.