Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiophenecarbonitrile, 97%

Catalog No. AC298050500
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC298050500 50 mL Glass bottle
AC298050100 10 mL Glass bottle
AC298052500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC298050500 Supplier Thermo Scientific Chemicals Supplier No. 298050500
Add to Cart

CAS: 1003-31-2 | C5H3NS | 109.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-31-2
Molecular Formula C5H3NS
Molecular Weight (g/mol) 109.15
MDL Number MFCD00005416
InChI Key CUPOOAWTRIURFT-UHFFFAOYSA-N
Synonym 2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile
PubChem CID 66087
IUPAC Name thiophene-2-carbonitrile
SMILES C1=CSC(=C1)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 192°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5620 to 1.564
Quantity 50 mL
Beilstein 18,29
Specific Gravity 1.17
Formula Weight 109.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Thiophenecarbonitrile, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
EINECSNumber 213-706-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.