Learn More
Apexbio Technology LLC Voglibose 83480-29-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B18705.1
Encompass_Preferred
Voglibose (CAS 83480-29-9) is an N-substituted derivative of valiolamine known for its inhibitory action on -glucosidase enzymes By selectively blocking these enzymes in the intestinal tract voglibose delays the breakdown and absorption of carbohydrates thereby attenuating postprandial hyperglycemic excursions This mechanism makes voglibose a valuable tool for investigating glucose metabolism and its related disorders including the pathophysiology of hyperglycemia and its associated complications Its use in preclinical and clinical research supports studies on the modulation of carbohydrate digestion and glycemic control
Catalog No. 50-000-89934
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.