Learn More
Apexbio Technology LLC Aspartame 22839-47-0 5g
SDP
Supplier:  Apexbio Technology LLC B18885000
Encompass_Preferred
Aspartame is a potent agonist targeting sweet taste receptors specifically G-protein coupled receptors (GPCRs) involved in gustatory signaling It is designed to activate these receptors thereby modulating taste perception and downstream signal transduction pathways Aspartame exerts its biological activity primarily through activation of sweet taste receptor-mediated signaling cascades In cellular and animal models Aspartame demonstrates agonist activity Based on these pharmacological properties Aspartame holds research potential in taste receptor studies investigation of receptor-mediated signaling pathways and exploration of metabolic and neurobiological processes related to amino acid metabolism and methanol processing
Catalog No. 50-000-92278
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.