Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone, 95%

Catalog No. AAH6344703
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6344703 1 g
AAH63447MD 250 mg
AAH6344706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6344703 Supplier Thermo Scientific Chemicals Supplier No. H6344703
Add to Cart

CAS: 315-08-2 | C10H6F3NOS2 | 277.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 315-08-2
Molecular Formula C10H6F3NOS2
Molecular Weight (g/mol) 277.28
MDL Number MFCD00126176
InChI Key MRDJXDJVVCNDND-UHFFFAOYSA-N
Synonym n-3-trifluoromethyl phenyl rhodanine, 2-thioxo-3-3-trifluoromethyl-phenyl-thiazolidin-4-one, 2-sulfanylidene-3-3-trifluoromethyl phenyl-1,3-thiazolidin-4-one, 2-thioxo-3-3-trifluoromethyl phenyl thiazolidin-4-one, 2-thioxo-3-3-trifluoromethyl phenyl-4-thiazolidinone, 2-thioxo-3-3-trifluoromethylhenyl-thiazolidin-4-one, 2-thioxo-3-3-trifluoromethyl phenyl-1,3-thiazolidin-4-one, 3-3-trifluoromethyl phenyl rhodanine, 2-tioxo-3-3-trifluoromethylphenyl thiazolidin-4-one
PubChem CID 278104
IUPAC Name 2-sulfanylidene-3-[3-(trifluoromethyl)phenyl]-1,3-thiazolidin-4-one
SMILES FC(F)(F)C1=CC=CC(=C1)N1C(=O)CSC1=S

Specifications

Color White
Quantity 1 g
Formula Weight 277.28
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.