Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethoxy)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. T23825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T23825G Supplier TCI America Supplier No. T23825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1979-29-9
Molecular Formula C8H4F3O3
Molecular Weight (g/mol) 205.11
MDL Number MFCD00052325
InChI Key JMYSPFGUBNENSE-UHFFFAOYSA-M
Synonym 2-trifluoromethoxy benzoic acid, o-trifluoromethoxybenzoic acid, alpha,alpha,alpha-trifluoro-o-anisic acid, 2-trifluoromethoxy-benzoic acid, rarechem al bo 0449, pubchem4587, acmc-209f2a, ksc494o7t
PubChem CID 2777223
IUPAC Name 2-(trifluoromethoxy)benzoate
SMILES [O-]C(=O)C1=CC=CC=C1OC(F)(F)F

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 5 g
Formula Weight 206.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethoxy)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.