Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethoxy)benzylamine 98.0+%, TCI America™
SDP

Catalog No. T30921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T30921G Supplier TCI America Supplier No. T30921G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 175205-64-8
Molecular Formula C8H9F3NO
Molecular Weight (g/mol) 192.16
MDL Number MFCD00085176
InChI Key FYXMMFFZMQMXCQ-UHFFFAOYSA-O
Synonym 2-trifluoromethoxy benzylamine, 2-trifluoromethoxy phenyl methanamine, 2-trifluoromethoxybenzylamine, o-trifluoromethoxybenzyl amine, 1-2-trifluoromethoxy phenyl methanamine, 2-trifluormethoxybenzylamine, benzenemethanamine, 2-trifluoromethoxy, 2-trifluoromethoxy phenyl methylamine, pubchem4432
PubChem CID 2777251
IUPAC Name [2-(trifluoromethoxy)phenyl]methanaminium
SMILES [NH3+]CC1=CC=CC=C1OC(F)(F)F

Specifications

Color Yellow
Boiling Point 77°C
Quantity 1 g
UN Number 2735
Formula Weight 191.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethoxy)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.