Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethyl)phenol, 98%

Catalog No. AAA1423106
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1423106 5 g
AAA1423103 1 g
AAA1423114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1423106 Supplier Thermo Scientific Chemicals Supplier No. A1423106
Add to Cart

CAS: 444-30-4 | C7H5F3O | 162.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444-30-4
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.111
MDL Number MFCD00002222
InChI Key ZOQOPXVJANRGJZ-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenol, 2-hydroxybenzotrifluoride, o-trifluoromethylphenol, o-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-o-cresol, phenol, 2-trifluoromethyl, o-trifluoromethoxy phenol, a,a,a-trifluoro-o-cresol, 2-hydroxy-alpha,alpha,alpha-trifluorotoluene
PubChem CID 67958
IUPAC Name 2-(trifluoromethyl)phenol
SMILES C1=CC=C(C(=C1)C(F)(F)F)O

Specifications

Melting Point 45°C to 46°C
Boiling Point 147°C to 148°C
Flash Point 65°C (149°F)
Odor Phenol-like
Quantity 5 g
UN Number UN3261
Beilstein 1867917
Formula Weight 162.11
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)phenol
Hazard Category H302+H312+H332-H318-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 207-148-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.