Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethyl)phenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. T166125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T166125G 25 g
T16611G 1 g
T16615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T166125G Supplier TCI America Supplier No. T166125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3038-48-0
Molecular Formula C9H6F3O2
Molecular Weight (g/mol) 203.14
MDL Number MFCD00004327
InChI Key TYOCDHCKTWANIR-UHFFFAOYSA-M
Synonym 2-trifluoromethyl phenylacetic acid, 2-2-trifluoromethyl phenyl acetic acid, 2-trifluoromethylphenylacetic acid, a,a,a-trifluoro-o-tolyl acetic acid, 2-trifluoromethyl phenyl acetic acid, alpha,alpha,alpha-trifluoro-o-tolyl acetic acid, benzeneacetic acid, 2-trifluoromethyl, acmc-209hfg, ksc494k8b
PubChem CID 594597
IUPAC Name 2-[2-(trifluoromethyl)phenyl]acetate
SMILES [O-]C(=O)CC1=CC=CC=C1C(F)(F)F

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 25 g
Formula Weight 204.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)phenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.