Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Trimethylsilyloxy-1,3-butadiene 95.0+%, TCI America™
SDP

Catalog No. T146925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T146925ML 25 mL
T14695ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T146925ML Supplier TCI America Supplier No. T146925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38053-91-7
Molecular Formula C7H14OSi
Molecular Weight (g/mol) 142.27
MDL Number MFCD00009852
InChI Key JOAPBVRQZQYKMS-UHFFFAOYSA-N
PubChem CID 554657
IUPAC Name (buta-1,3-dien-2-yloxy)trimethylsilane
SMILES C[Si](C)(C)OC(=C)C=C

Specifications

Color Yellow
Boiling Point 55°C
Quantity 25 mL
UN Number 1993
Formula Weight 142.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Trimethylsilyloxy-1,3-butadiene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.