Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trimethylsilyloxy)furan 96.0+%, TCI America™
SDP

Catalog No. T153625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T153625G 25 g
T15365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T153625G Supplier TCI America Supplier No. T153625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61550-02-5
Molecular Formula C7H12O2Si
Molecular Weight (g/mol) 156.26
MDL Number MFCD00011729
InChI Key ILBCDLOQMRDXLN-UHFFFAOYSA-N
Synonym (2-Furanyloxy)trimethylsilane
PubChem CID 188414
IUPAC Name (furan-2-yloxy)trimethylsilane
SMILES C[Si](C)(C)OC1=CC=CO1

Specifications

Color Yellow
Boiling Point 50°C
Quantity 25 g
UN Number 1993
Formula Weight 156.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilyloxy)furan
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.