Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Vinylanthraquinone 98.0+%, TCI America™
SDP

Catalog No. V01391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01391G 1 g
V01395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01391G Supplier TCI America Supplier No. V01391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13388-33-5
Molecular Formula C16H10O2
Molecular Weight (g/mol) 234.25
MDL Number MFCD18448904
InChI Key IQOUOAIZDKROQT-UHFFFAOYSA-N
PubChem CID 22708978
IUPAC Name 2-ethenyl-9,10-dihydroanthracene-9,10-dione
SMILES C=CC1=CC2=C(C=C1)C(=O)C1=CC=CC=C1C2=O

Specifications

Melting Point 177°C
Color Yellow
Quantity 1 g
Formula Weight 234.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Vinylanthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.