Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Vinylpyrazine, 98%, stab. with ca 0.1% hydroquinone

Catalog No. AAA1400703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1400703 1 g
AAA1400706 5 g
AAA1400714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1400703 Supplier Thermo Scientific Chemicals Supplier No. A1400703
Add to Cart

CAS: 4177-16-6 | C6H6N2 | 106.13 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Slightly soluble in methanol, water.

Notes
Store away from incompatible compounds such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4177-16-6
Molecular Formula C6H6N2
Molecular Weight (g/mol) 106.13
MDL Number MFCD00051591
InChI Key KANZWHBYRHQMKZ-UHFFFAOYSA-N
Synonym 2-vinylpyrazine, vinylpyrazine, pyrazine, ethenyl, pyrazine, vinyl, 2-vinyl pyrazine, unii-y6tpk237o6, ethenylpyrazine, vinyl pyrazine, 2ethenylpyrazine, acmc-20anmb
PubChem CID 77840
IUPAC Name 2-ethenylpyrazine
SMILES C=CC1=CN=CC=N1

Specifications

Density 1.041
Boiling Point 163°C to 164°C
Flash Point 60°C (140°F)
Refractive Index 1.556
Quantity 1 g
UN Number UN1993
Beilstein 105566
Solubility Information Slightly soluble in methanol,water.
Formula Weight 106.13
Percent Purity 98%
Chemical Name or Material 2-Vinylpyrazine, stabilized with ≈0.1% hydroquinone
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 224-045-2
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.