Learn More
National Safety Apparel™ 20 Cal Ultrasoft Arc Flash Coat
SDP
Catalog No. 17001272 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-272 2X-Large
17-001-269 Small
17-001-270 Medium
17-001-271 Large
17-001-273 3X-Large
17-001-274 X-Large
17-001-275 4X-Large
17-001-276 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-272 Supplier National Safety Apparel™ Supplier No. C04UW032X32
Add to Cart
Add to Cart
For enhanced protection Westex™ Ultrasoft is the ideal choice for superior comfort, improved safety, and above average durability
  • Arc Rating: 20 cal/cm²
  • CAT 2
  • Fabric/Material: 13 oz. Westex™ Ultrasoft
  • Breathable FR fabric
  • FR fabric will not melt, drip or ignite
  • FR knit wrist cuffs keep sleeves in place
  • Generous fit for ease of movement
  • Inherently flame resistant protection will not wash/wear out
  • Hook and loop front closure
  • Made in the USA
Specifications
Closure Type Hook and Loop
Color Navy
Disposable Reusable
Gender Unisex
Material Cotton, Nylon
Neck Style Collared
Size 2X-Large
Resistance Flame
Seam Type Stitched
Arc Flash Rating 20 cal/cm²
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, CSA Z462