Learn More
Cayman Chemical 20-hydroxy ArachIdIc AcId 25mg
SDP
Supplier:  Cayman Chemical 2465225 MG
Encompass_Preferred
A hydroxy fatty acid; has been found in silver birch (B. pendula) as well as Q. ilex from Cala Violina and Colognole, Italy; identified as a potential inhibitor of human rhinovirus coat protein in a structure-based virtual screen
Catalog No. 50-293-2492