Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

20-Hydroxyecdysone, ≥95%, MP Biomedicals™

Catalog No. ICN15743410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15743410 10 mg
ICN15743401 1 mg
ICN15743405 5 mg
ICN15743425 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15743410 Supplier MP Biomedicals, Inc Supplier No. 0215743410
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5289-74-7
Molecular Formula C27H44O7
Molecular Weight (g/mol) 480.64
MDL Number MFCD00036740
InChI Key NKDFYOWSKOHCCO-YPVLXUMRSA-N
Synonym 20-hydroxyecdysone, ecdysterone, beta-ecdysone, crustecdysone, 20-oh ecdysone, the-7, polypodine a, insect moulting hormone, commisterone, crustecdyson
PubChem CID 5459840
ChEBI CHEBI:16587
SMILES CC(C)(O)CC[C@@H](O)[C@](C)(O)[C@H]1CC[C@@]2(O)C3=CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@H]3CC[C@]12C

Specifications

Assay Percent Range ≥95%
Quantity 10 mg
IUPAC Name (1S,3aS,5aR,7R,8S,9aR,9bR,11aR)-3a,7,8-trihydroxy-9a,11a-dimethyl-1-[(2R,3R)-2,3,6-trihydroxy-6-methylheptan-2-yl]-1H,2H,3H,3aH,5H,5aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-5-one
Formula Weight 480.6
Physical Form Powder
Chemical Name or Material 20-Hydroxyecdysone
Recommended Storage Store at 0°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.