Learn More
Eisco Labs 20 L HDPE Jerrican with Silicone Lined Polypropylene Cap
SCI_ED
Catalog No. 1675235 Shop All Eisco Products
Change view
Click to view available options
Color:
Translucent
Catalog No. Color
16-752-35 Translucent
1 options
Catalog No. 16-752-35 Supplier Eisco Labs Supplier No. CH5592
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
20 L HDPE jerrican with wide-mouth opening and ergonomic handle that is FDA and USP compliant, leak-tested, and built for safe liquid storage, mixing, and transport in lab or industrial settings across a wide temperature range.
  • Durable HDPE Construction: Made from HDPE, this 20 L jerrican is designed for reliable liquid storage, mixing, and transportation in laboratory and industrial settings.
  • Wide Temperature Range: Handles temperatures from -100°C to 120°C, offering versatility for a wide range of scientific and industrial liquid handling needs.
  • User-Friendly Design: Features an ergonomic handle for easy transport and a wide-mouth opening for convenient filling and cleaning, ensuring a seamless user experience.
  • Certified Materials: FDA-compliant (21 CFR 177.1520), USP 88 Class VI biocompatible, and BSE/TSE free, suitable for regulated and sensitive substances.
  • Leakproof Tested: Rigorously tested under IS 6312 standards for crack resistance and hydrostatic strength, ensuring safe and efficient liquid storage and transportation.
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 20 L
Color Translucent
Material HDPE
Mouth Style Narrow
Product Type Jerry Can
Sterility Non-sterile