Description
- For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
- Reduces the number of locks being used
- Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
- Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
| Color | Black on Yellow |
| Displayed Text | LOCKOUT STATION |
| Includes | 20 in. ''Estacion de Bloqueo'' Board, 20 ''Peligro No Operar'' Tags |
| Quantity | 1 Kit |