Learn More
Heathrow Scientific™ 20-Place Cardboard Slide Tray
GREENER_CHOICE
Catalog No. 17100417
Change view
Click to view available options
Color:
Blue
Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-100-417 Blue
17-100-442 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-100-417 Supplier Heathrow Scientific™ Supplier No. HS9913
Add to Cart
Add to Cart
20-Place Cardboard Slide Tray holds standard slides.
  • Index on the front cover corresponds with the number grid inside for identification
  • Recessed slide compartments and inside covers for specimen protection
  • Slides cannot be accessed when cover is closed reducing contamination
  • Compartment holder is made from a double layer of densely pressed cardboard
  • Cover is single layer cardboard
  • Front protection flaps are attached with durable tape
Specifications
Type Slide Holder
Slide Capacity 20
Color Blue
Dimensions (L x W x H) 8.2 x 13.4 x 0.4 in. (20.8 x 34.1 x 0.9 cm)