Learn More
Cayman Chemical 20 R-ProtopnaxatrIol 25mg
SDP
Supplier:  Cayman Chemical 3653025 MG
Encompass_Preferred
A coumarin glucoside inhibits A23187-induced production of PGE2 and LTC4 in isolated mouse peritoneal macrophages at 100 UM reduces synovial inflammation and fibrosis as well as bone and cartilage erosion in the inflamed joints in a complete Freunds adjuvant- and M butyricum-induced rat model of rheumatoid arthritis at 50 and 100 mg/kg
Catalog No. 50-291-2323
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.