Learn More
ATCC 20 strain mix; whole cell material
Supplier:  ATCC MSA2002
Encompass
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product is prepared as a mixture of whole cells and comprises fully sequenced, characterized, and authenticated ATCC Genuine Cultures that were selected based on relevant phenotypic and genotypic attributes, such as Gram stain, GC content, genome size, and spore formation. Each order for this product is provided with access to run 5 analyses on One Codex, the leading bioinformatics platform for microbial genomics and metagenomics. Specification Range: 4x107 cells/vial ± 1 log* *Whole cell concentrations indicate ATCC manufacturing specifications and are provided as a reference only
Catalog No. 50-238-4928
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.