Learn More
Bioassay Systems Urinary Indican Test Kit. The urinary Indican test is an accurate, simple, fast (15 min) and inexpensive screening tool for the identification of protein putrefaction in the gastrointestinal tract.
SDP
Catalog No. 501956329 Shop All Bioassay Systems Products
Encompass_Preferred
An accurate, simple, fast (15 min) and inexpensive screening tool for the identification of protein putrefaction in the gastrointestinal tract. For research use only, not intended for diagnostic use. The Indican Reagent is corrosive. It is recommended to perform the test in a chemical fume hood. Wear gloves, goggles and protective clothing. Key Features: Convenient. Only need to pipette 2 mL urine into the ready reagent vial, mix and read the indican level from a color chart. Fast: 15 min. Method: Obermeyer (Improved). Samples: Urine. Species: Human. Procedure: Assay takes 15 min. Kit size: 20 tests.
Catalog No. 50-195-6329 Supplier Bioassay Systems Supplier No. I1000N
An accurate, simple, fast (15 min) and inexpensive screening tool for the identification of protein putrefaction in the gastrointestinal tract. For research use only, not intended for diagnostic use. The Indican Reagent is corrosive. It is recommended to perform the test in a chemical fume hood. Wear gloves, goggles and protective clothing. Key Features: Convenient. Only need to pipette 2 mL urine into the ready reagent vial, mix and read the indican level from a color chart. Fast: 15 min. Method: Obermeyer (Improved). Samples: Urine. Species: Human. Procedure: Assay takes 15 min. Kit size: 20 tests.