Learn More
POLYCARBIN INC 20 ul Universal Fit Pipette Tips (Non-Filtered) CS50
SDP
Supplier:  POLYCARBIN INC PC010604
Encompass_Preferred
Polycarbin's pipette tip rack is made of 100% circular economy, post-consumer recycled content. Pipette tips do not contain recycled content, and are universal fit, sterile, and certified DNase, RNase, and Pyrogen free. Racked: 96 Tips/Rack; 10 Racks/Pack; 5 Packs/Case. Manufactured in an ISO 9001 compliant facility.
Catalog No. NC2210206
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.