Learn More
Description
- Meets USP specifications
- Manufactured in ISO 9001 certified facility
- Lot-specific certificate of analysis
- For production
- Includes 2-port top drain
Applications:
- Hydration of cell culture media, supplements and salt solution
- Rinsing of primary packaging, production vessels, equipment and room surfaces
- Reconsitution and rehydration of product during synthesis
- Preparation of rinse, purification and chromatography buffer
Specifications
Specifications
| Color | Clear |
| Conductivity | ≤ 5 μS/cm at 24 to 26 °C |
| Endotoxin | < 0.25 EU/mL |
| Grade | EP/USP |
| Sterility | Sterile |
| Quantity | 200 L |
| Chemical Name or Material | Water |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.