Learn More
Fisherbrand™ 200 Series Safety Glasses
SCI_ED
Economical, lightweight protection.
Specifications
Lens Coating Hardcoat
Certifications/Compliance CSA approved; Meet ANSI Z87.1-2003 standards.
Lens Tint Clear
Temple Style Adjustable; spatula
View More Specs
Products 2
Catalog Number Frame Color Price Quantity & Availability  
Catalog Number Frame Color Price Quantity & Availability  
S01822
SCI_ED
View Documents
Black N/A N/A N/A
S01821
SCI_ED
View Documents
Blue N/A N/A N/A
Description
Description
  • One-piece lens provides a full and unobstructed field of vision
  • Absorb >99.9% of harmful UV light
  • Molded-in sideshields and browguards
  • Adjustable spatula temples
Certifications
CSA-approved.
Compliance
Meet ANSI™ Z87.1-2003 standards.
Specifications
Specifications
Hardcoat
CSA approved; Meet ANSI Z87.1-2003 standards.
Clear
Adjustable; spatula
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.