Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2000 μg/mL 8260 MegaMix Calibration Mix, Revised (75 components) in P&T Methanol, Restek
SDP

Catalog No. 06611988 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-611-988 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-611-988 Supplier Restek Corporation Supplier No. 30152
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For analysis of VOCs in solid waste

  • Quantitatively analyzed to confirm concentration and identity of each compound
  • Excludes pentachloroethane for labs experiencing elevated tetrachloroethene response caused by pentachloroethane breakdown

Specifications

Chemical Name or Material 8260 MegaMix Calibration Mix, Revised
Solution Contains 75 components
Shipping Condition Ambient
Concentration 2000 μg/mL
Certifications/Compliance Certified reference material
Solvent or Matrix Methanol-P and T
Industry Type Solid Waste Testing
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.