Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2000 μg/mL EPA 524.3 Reference Standards (5 Mixes Including MegaMix Standard) in P&T Methanol, Restek
SDP

Catalog No. 06611985 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-611-985 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-611-985 Supplier Restek Corporation Supplier No. 30013
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Can be used for surface water and groundwater testing

  • Full 82-component EPA 524.3 list using as few as three ampuls
    • Reduces prep time and chances for error or contamination
  • EPA 524.3 VOA MegaMix ampul includes oxygenates group
    • No need to order separately
  • Volatile gases prepared separately
    • Replaces shorter life components without wasting money on full list
  • Two options for internal and surrogate standards
    • Separate or combined mix

Specifications

Shipping Condition Ambient
Concentration 2000 μg/mL
Certifications/Compliance Certified reference material
Solvent or Matrix Methanol-P and T
Industry Type Surface Water Testing, Groundwater Testing
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.